Methyl nitroacetate 98%

BD01387608-100 BD01387608-25G
BLDPBD01387608-100EA 574 EUR
BLDPBD01387608-100 BLDPBD01387608-25G
Methyl nitroacetate 98%
Methyl nitroacetate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₃H₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD00075480
CAS Number: 2483-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR