1-(2-Tetrahydropyranyl)-1H-pyrazole 98+%
|
|
BD01392245-500
BD01392245-100
BLDPBD01392245-500EA
386
EUR
BLDPBD01392245-500
BLDPBD01392245-100
1-(2-Tetrahydropyranyl)-1H-pyrazole 98+%
1-(2-Tetrahydropyranyl)-1H-pyrazole
Storage condition: Sealed in dry, Room Temperature
Formula:
C₈H₁₂N₂O Storage Temperature: Ambient |
MDL Number:
MFCD11044692 CAS Number: 449758-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...