3-Chloro-4-(trifluoromethyl)benzonitrile 98%

BD01393461-1G BD01393461-25G
BLDPBD01393461-1GEA 9.5 EUR
BLDPBD01393461-1G BLDPBD01393461-25G
3-Chloro-4-(trifluoromethyl)benzonitrile 98%
3-Chloro-4-(trifluoromethyl)benzonitrile

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₃ClF₃N
Storage Temperature: Ambient
MDL Number: MFCD04972703
CAS Number: 1092460-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR