(23S,50S)-50-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-23-(tert-butoxycarbonyl)-2,2-dimethyl-4,21,26,35,44-pentaoxo-3,30,33,39,42-pentaoxa-22,27,36,45-tetraazahenpentacontan-51-oic acid 98%

BD01393810-100 BD01393810-1G
BLDPBD01393810-100EA 96.6 EUR
BLDPBD01393810-100 BLDPBD01393810-1G
(23S,50S)-50-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-23-(tert-butoxycarbonyl)-2,2-dimethyl-4,21,26,35,44-pentaoxo-3,30,33,39,42-pentaoxa-22,27,36,45-tetraazahenpentacontan-51-oic acid 98%
(23S,50S)-50-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-23-(tert-butoxycarbonyl)-2,2-dimethyl-4,21,26,35,44-pentaoxo-3,30,33,39,42-pentaoxa-22,27,36,45-tetraazahenpentacontan-51-oic acid

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formula: C₆₄H₁₀₁N₅O₁₆
Storage Temperature: Freezer
CAS Number: 1662688-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR