1-(Bromomethyl)-4-isopropylbenzene 96%
|
|
BD01400132-5G
BD01400132-1G
BLDPBD01400132-5GEA
27.3
EUR
BLDPBD01400132-5G
BLDPBD01400132-1G
1-(Bromomethyl)-4-isopropylbenzene 96%
1-(Bromomethyl)-4-isopropylbenzene
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₁₀H₁₃Br MW: 213.118 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03427264 CAS Number: 73789-86-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...