1-Methyl-[4,4'-bipyridine]-1,1'-diium chloride 95%

BD01406253-250 BD01406253-1G
BLDPBD01406253-250EA 75.6 EUR
BLDPBD01406253-250 BLDPBD01406253-1G
1-Methyl-[4,4'-bipyridine]-1,1'-diium chloride 95%
1-Methyl-[4,4'-bipyridine]-1,1'-diium chloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₁H₁₂Cl₂N₂
MW: 243.1324 g/mol
Storage Temperature: Ambient
CAS Number: 4397-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR