Ethyl 1-(3-chlorophenyl)-1H-imidazole-5-carboxylate 95%

BD01409178-5G BD01409178-100
BLDPBD01409178-5GEA 244 EUR
BLDPBD01409178-5G BLDPBD01409178-100
Ethyl 1-(3-chlorophenyl)-1H-imidazole-5-carboxylate 95%
Ethyl 1-(3-chlorophenyl)-1H-imidazole-5-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₁₁ClN₂O₂
Storage Temperature: Ambient
CAS Number: 2110819-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR