Fmoc-3-amino-3-phenylpropionic acid 98+%

BD01411518-5G BD01411518-1G
BLDPBD01411518-5GEA 252 EUR
BLDPBD01411518-5G BLDPBD01411518-1G
Fmoc-3-amino-3-phenylpropionic acid 98+%
Fmoc-3-amino-3-phenylpropionic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₄H₂₁NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD01863192
CAS Number: 180181-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR