trans-2-(3-Bromophenyl)cyclopropan-1-amine hydrochloride 98%

BD01416582-100 BD01416582-250
BLDPBD01416582-100EA 108 EUR
BLDPBD01416582-100 BLDPBD01416582-250
trans-2-(3-Bromophenyl)cyclopropan-1-amine hydrochloride 98%
trans-2-(3-Bromophenyl)cyclopropan-1-amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₁₁BrClN
MW: 248.5473 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28118779
CAS Number: 1807920-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR