4,4'-(1,4-Phenylenebis(ethyne-2,1-diyl))dianiline 95%

BD01418222-250 BD01418222-100
BLDPBD01418222-250EA 115 EUR
BLDPBD01418222-250 BLDPBD01418222-100
4,4'-(1,4-Phenylenebis(ethyne-2,1-diyl))dianiline 95%
4,4'-(1,4-Phenylenebis(ethyne-2,1-diyl))dianiline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₂₂H₁₆N₂
Storage Temperature: Ambient
CAS Number: 246141-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR