11-(tert-Butoxy)-11-oxoundecanoic acid 98+%

BD01432700-100 BD01432700-25G
BLDPBD01432700-100EA 686 EUR
BLDPBD01432700-100 BLDPBD01432700-25G
11-(tert-Butoxy)-11-oxoundecanoic acid 98+%
11-(tert-Butoxy)-11-oxoundecanoic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₅H₂₈O₄
MW: 272.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30737824
CAS Number: 1789702-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR