4,4'-Di(1H-1,2,4-triazol-1-yl)-1,1'-biphenyl 95%
|
|
BD01438728-250
BD01438728-1G
BLDPBD01438728-250EA
140
EUR
BLDPBD01438728-250
BLDPBD01438728-1G
4,4'-Di(1H-1,2,4-triazol-1-yl)-1,1'-biphenyl 95%
4,4'-Di(1H-1,2,4-triazol-1-yl)-1,1'-biphenyl
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₆H₁₂N₆ Storage Temperature: Ambient |
CAS Number:
1642849-37-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...