2-(4-((3-Chlorobenzyl)oxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
|
|
BD01456059-5G
BD01456059-250
BLDPBD01456059-5GEA
648
EUR
BLDPBD01456059-5G
BLDPBD01456059-250
2-(4-((3-Chlorobenzyl)oxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(4-((3-Chlorobenzyl)oxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Storage condition: Sealed in dry, Room Temperature
Formula:
C₂₁H₂₆BClO₃ Storage Temperature: Ambient |
CAS Number:
2828440-10-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...