1-Tetradecyl-3-methylimidazolium chloride 95%
|
|
BD01462360-5G
BD01462360-100
BLDPBD01462360-5GEA
8.4
EUR
BLDPBD01462360-5G
BLDPBD01462360-100
1-Tetradecyl-3-methylimidazolium chloride 95%
1-Tetradecyl-3-methylimidazolium chloride
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₈H₃₅ClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD03427607 CAS Number: 171058-21-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...