2-Amino-7-ethyl-5-oxo-5H-chromeno[2,3-b]pyridine-3-carboxylic acid 95%

BD01513065-50M BD01513065-25M
BLDPBD01513065-50MEA 473 EUR
BLDPBD01513065-50M BLDPBD01513065-25M
2-Amino-7-ethyl-5-oxo-5H-chromeno[2,3-b]pyridine-3-carboxylic acid 95%
2-Amino-7-ethyl-5-oxo-5H-chromeno[2,3-b]pyridine-3-carboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₅H₁₂N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD06208374
CAS Number: 68302-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR