6-Methoxyquinaldine 98%

BD0511-10G BD0511-1G
BLDPBD0511-10GEA 52.5 EUR
BLDPBD0511-10G BLDPBD0511-1G
6-Methoxyquinaldine 98%
6-Methoxyquinaldine

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD00006761
CAS Number: 1078-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR