5-Methoxy-2-tetralone 95%

BD0541-1G BD0541-25G
BLDPBD0541-1GEA 13.7 EUR
BLDPBD0541-1G BLDPBD0541-25G
5-Methoxy-2-tetralone 95%
5-Methoxy-2-tetralone

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₁₁H₁₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00064958
CAS Number: 32940-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR