N-Ethyl-N-(3-sulphopropyl)-M-anisidine sodium 97%
|
|
BD0926-1G
BD0926-250MG
BLDPBD0926-1GEA
30.5
EUR
BLDPBD0926-1G
BLDPBD0926-250MG
N-Ethyl-N-(3-sulphopropyl)-M-anisidine sodium 97%
N-Ethyl-N-(3-sulphopropyl)-m-anisidine sodium
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₁₂H₁₈NNaO₄S MW: 295.3304 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00083320 CAS Number: 82611-88-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...