Ethyl-4-pyrazolecarboxylate 98%

BD10016-100G BD10016-25G
BLDPBD10016-100GEA 123 EUR
BLDPBD10016-100G BLDPBD10016-25G
Ethyl-4-pyrazolecarboxylate 98%
Ethyl-4-pyrazolecarboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00010844
CAS Number: 37622-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR