Ethyl 4-(trifluoromethyl)thiazole-2-carboxylate 95%

BD100500-1G BD100500-5G BD100500-100MG
BLDPBD100500-1GEA 125 EUR
BLDPBD100500-1G BLDPBD100500-5G BLDPBD100500-100MG
Ethyl 4-(trifluoromethyl)thiazole-2-carboxylate 95%
Ethyl 4-(trifluoromethyl)thiazole-2-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₆F₃NO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD08669898
CAS Number: 79247-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR