L-Glutamic acid 5-ethyl ester 97%

BD10090-100G BD10090-1G
BLDPBD10090-100GEA 123 EUR
BLDPBD10090-100G BLDPBD10090-1G
L-Glutamic acid 5-ethyl ester 97%
L-Glutamic acid 5-ethyl ester

Storage condition: Keep in dark place, Sealed in dry, Store in freezer, under -20°C

Formula: C₇H₁₃NO₄
Storage Temperature: Freezer
MDL Number: MFCD00037217
CAS Number: 1119-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR