2-Bromo-α,α,α,4-tetrafluorotoluene 98%
|
|
BD10149-25G
BD10149-500G
BLDPBD10149-25GEA
68.3
EUR
BLDPBD10149-25G
BLDPBD10149-500G
2-Bromo-α,α,α,4-tetrafluorotoluene 98%
2-Bromo-α,α,α,4-tetrafluorotoluene
Storage condition: Sealed in dry, Room Temperature
Formula:
C₇H₃BrF₄ Storage Temperature: Ambient |
MDL Number:
MFCD03094041 CAS Number: 351003-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...