2-Bromo-α,α,α,5-tetrafluorotoluene 98%

BD10150-10G BD10150-500G
BLDPBD10150-10GEA 9.5 EUR
BLDPBD10150-10G BLDPBD10150-500G
2-Bromo-α,α,α,5-tetrafluorotoluene 98%
2-Bromo-α,α,α,5-tetrafluorotoluene

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₃BrF₄
Storage Temperature: Ambient
MDL Number: MFCD00040937
CAS Number: 40161-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR