5-Bromo-2-fluoro-m-xylene 98%

BD102718-25G BD102718-10G
BLDPBD102718-25GEA 102 EUR
BLDPBD102718-25G BLDPBD102718-10G
5-Bromo-2-fluoro-m-xylene 98%
5-Bromo-2-fluoro-m-xylene

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₈BrF
MW: 203.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01320701
CAS Number: 99725-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR