5-Bromo-2-methyl-4-nitro-1H-imidazole 95%

BD102801-1G BD102801-25G
BLDPBD102801-1GEA 26.3 EUR
BLDPBD102801-1G BLDPBD102801-25G
5-Bromo-2-methyl-4-nitro-1H-imidazole 95%
5-Bromo-2-methyl-4-nitro-1H-imidazole

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₄H₄BrN₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00156130
CAS Number: 18874-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR