4-Bromo-2-(trifluoromethoxy)aniline 98%

BD10330-10G BD10330-5G
BLDPBD10330-10GEA 10.5 EUR
BLDPBD10330-10G BLDPBD10330-5G
4-Bromo-2-(trifluoromethoxy)aniline 98%
4-Bromo-2-(trifluoromethoxy)aniline

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₇H₅BrF₃NO
Storage Temperature: Refrigerator
MDL Number: MFCD00179338
CAS Number: 175278-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR