2-Fluoro-6-iodotoluene 98%

BD10455-5G BD10455-10G
BLDPBD10455-5GEA 15.8 EUR
BLDPBD10455-5G BLDPBD10455-10G
2-Fluoro-6-iodotoluene 98%
2-Fluoro-6-iodotoluene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₆FI
Storage Temperature: Ambient
MDL Number: MFCD00142922
CAS Number: 443-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR