5-Methyl-1,3,4-oxadiazol-2-amine 98%

BD104620-25G BD104620-10G
BLDPBD104620-25GEA 251 EUR
BLDPBD104620-25G BLDPBD104620-10G
5-Methyl-1,3,4-oxadiazol-2-amine 98%
5-Methyl-1,3,4-oxadiazol-2-amine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₃H₅N₃O
Storage Temperature: Refrigerator
MDL Number: MFCD03703470
CAS Number: 52838-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR