5-Nitro-1,3-dihydro-2H-benzimidazol-2-one 97%

BD104988-5G BD104988-10G
BLDPBD104988-5GEA 38.9 EUR
BLDPBD104988-5G BLDPBD104988-10G
5-Nitro-1,3-dihydro-2H-benzimidazol-2-one 97%
5-Nitro-1,3-dihydro-2H-benzimidazol-2-one

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₅N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD00220274
CAS Number: 93-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR