5-Nitrobenzo[d]thiazole 97%

BD105032-1G BD105032-100MG
BLDPBD105032-1GEA 38.9 EUR
BLDPBD105032-1G BLDPBD105032-100MG
5-Nitrobenzo[d]thiazole 97%
5-Nitrobenzo[d]thiazole

Storage condition: Sealed in dry, Room Temperature

Formula: C7H4N2O2S
Storage Temperature: Ambient
MDL Number: MFCD00085891
CAS Number: 2942-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR