5-Nitro-2-thiophenecarbonitrile 98%

BD105083-5G BD105083-10G
BLDPBD105083-5GEA 30.5 EUR
BLDPBD105083-5G BLDPBD105083-10G
5-Nitro-2-thiophenecarbonitrile 98%
5-Nitro-2-thiophenecarbonitrile

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₅H₂N₂O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD00052401
CAS Number: 16689-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR