3-Nitro-p-cresol 98%

BD10533-10G BD10533-25G
BLDPBD10533-10GEA 10.5 EUR
BLDPBD10533-10G BLDPBD10533-25G
3-Nitro-p-cresol 98%
3-Nitro-p-cresol

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD00007244
CAS Number: 2042-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR