N(α)-Acetyl-D-tryptophan 98%

BD105853-100G BD105853-25G
BLDPBD105853-100GEA 351 EUR
BLDPBD105853-100G BLDPBD105853-25G
N(α)-Acetyl-D-tryptophan 98%
N(α)-Acetyl-D-tryptophan

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₃H₁₄N₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00065021
CAS Number: 2280-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR