1,5-Naphthyridin-4-ol 95%

BD106229-5G BD106229-250MG
BLDPBD106229-5GEA 401 EUR
BLDPBD106229-5G BLDPBD106229-250MG
1,5-Naphthyridin-4-ol 95%
1,5-Naphthyridin-4-ol

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD00234402
CAS Number: 5423-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR