Acyclovir 98%

BD10675-25G BD10675-100G
BLDPBD10675-25GEA 27.3 EUR
BLDPBD10675-25G BLDPBD10675-100G
Acyclovir 98%
Acyclovir

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₈H₁₁N₅O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00057880
CAS Number: 59277-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR