Monosodium 5-sulphoisophthalate 97%
|
|
BD107381-100G
BD107381-25G
BLDPBD107381-100GEA
7.4
EUR
BLDPBD107381-100G
BLDPBD107381-25G
Monosodium 5-sulphoisophthalate 97%
Monosodium 5-sulphoisophthalate
Storage condition: Sealed in dry, Room Temperature
Formula:
C₈H₆O₇S Storage Temperature: Ambient |
MDL Number:
MFCD00007495 CAS Number: 6362-79-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...