N-Boc-6-Bromohexylamine 97%

BD107666-5G BD107666-1G
BLDPBD107666-5GEA 176 EUR
BLDPBD107666-5G BLDPBD107666-1G
N-Boc-6-Bromohexylamine 97%
N-Boc-6-Bromohexylamine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₁H₂₂BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06201019
CAS Number: 142356-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR