6-(Bromomethyl)-2-methylquinoline 98%

BD107670-5G BD107670-1G
BLDPBD107670-5GEA 692 EUR
BLDPBD107670-5G BLDPBD107670-1G
6-(Bromomethyl)-2-methylquinoline 98%
6-(Bromomethyl)-2-methylquinoline

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₁H₁₀BrN
Storage Temperature: Refrigerator
MDL Number: MFCD06658974
CAS Number: 141848-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR