6-Aminopyridazin-3(2H)-one 97%
|
|
BD108098-5G
BD108098-25G
BD108098-250MG
BLDPBD108098-5GEA
365
EUR
BLDPBD108098-5G
BLDPBD108098-25G
BLDPBD108098-250MG
6-Aminopyridazin-3(2H)-one 97%
6-Aminopyridazin-3(2H)-one
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₄H₅N₃O Storage Temperature: Ambient |
MDL Number:
MFCD11520599 CAS Number: 57041-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...