3,5-Bis(trifluoromethyl)phenol 97%

BD10848-100G BD10848-25G
BLDPBD10848-100GEA 118 EUR
BLDPBD10848-100G BLDPBD10848-25G
3,5-Bis(trifluoromethyl)phenol 97%
3,5-Bis(trifluoromethyl)phenol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₄F₆O
MW: 230.11 g/mol
Boiling Pt: 176
Melting Pt: 21
Storage Temperature: Ambient
MDL Number: MFCD00000386
CAS Number: 349-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR