6-Chloro-4-(trifluoromethyl)nicotinic acid 95%

BD108827-1G BD108827-100MG
BLDPBD108827-1GEA 125 EUR
BLDPBD108827-1G BLDPBD108827-100MG
6-Chloro-4-(trifluoromethyl)nicotinic acid 95%
6-Chloro-4-(trifluoromethyl)nicotinic acid

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₃ClF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD06800446
CAS Number: 261635-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR