3',5'-Dichloroacetophenone 97%

BD10943-5G BD10943-25G
BLDPBD10943-5GEA 32.6 EUR
BLDPBD10943-5G BLDPBD10943-25G
3',5'-Dichloroacetophenone 97%
3',5'-Dichloroacetophenone

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₆Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD00045189
CAS Number: 14401-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR