2-Fluoro-m-toluonitrile 95%

BD10970-1G BD10970-10G
BLDPBD10970-1GEA 11.6 EUR
BLDPBD10970-1G BLDPBD10970-10G
2-Fluoro-m-toluonitrile 95%
2-Fluoro-m-toluonitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₆FN
Storage Temperature: Ambient
MDL Number: MFCD07782078
CAS Number: 185147-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR