6-Methylpyrazine-2-carboxylic acid 97%

BD109769-250MG BD109769-10G
BLDPBD109769-250MGEA 24.2 EUR
BLDPBD109769-250MG BLDPBD109769-10G
6-Methylpyrazine-2-carboxylic acid 97%
6-Methylpyrazine-2-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₆H₆N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08705756
CAS Number: 5521-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR