4-Chloro-2-methoxybenzoic acid 98%

BD11033-100G BD11033-500G
BLDPBD11033-100GEA 102 EUR
BLDPBD11033-100G BLDPBD11033-500G
4-Chloro-2-methoxybenzoic acid 98%
4-Chloro-2-methoxybenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₇ClO₃
Storage Temperature: Ambient
MDL Number: MFCD00002532
CAS Number: 57479-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR