7-Bromo-4-chloro-2-methylquinazoline 97%
|
|
BD110357-5G
BD110357-100MG
BD110357-10G
BLDPBD110357-5GEA
225
EUR
BLDPBD110357-5G
BLDPBD110357-100MG
BLDPBD110357-10G
7-Bromo-4-chloro-2-methylquinazoline 97%
7-Bromo-4-chloro-2-methylquinazoline
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₉H₆BrClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD09746300 CAS Number: 403850-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...