3-(4-Pyridyl)acrylic acid 98%
|
|
BD11080-1G
BD11080-100G
BLDPBD11080-1GEA
7.4
EUR
BLDPBD11080-1G
BLDPBD11080-100G
3-(4-Pyridyl)acrylic acid 98%
3-(4-Pyridyl)acrylic acid
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₈H₇NO₂ MW: 149.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00040746 CAS Number: 5337-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...