Methyl-2-amino-3-chlorobenzoate 97%

BD11122-10G BD11122-500G
BLDPBD11122-10GEA 10.5 EUR
BLDPBD11122-10G BLDPBD11122-500G
Methyl-2-amino-3-chlorobenzoate 97%
Methyl 2-amino-3-chlorobenzoate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₈H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD06797833
CAS Number: 77820-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR