4-Amino-L-Phenylalanine 95%

BD11321-5G BD11321-25G
BLDPBD11321-5GEA 47.3 EUR
BLDPBD11321-5G BLDPBD11321-25G
4-Amino-L-Phenylalanine 95%
4-Amino-L-phenylalanine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₁₂N₂O₂
MW: 180.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00069927
CAS Number: 943-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR