4-((3-Hydroxypropyl)amino)-3-nitrophenol 95%

BD11331-100G BD11331-25G
BLDPBD11331-100GEA 119 EUR
BLDPBD11331-100G BLDPBD11331-25G
4-((3-Hydroxypropyl)amino)-3-nitrophenol 95%
4-((3-Hydroxypropyl)amino)-3-nitrophenol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₁₂N₂O₄
MW: 212.2000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368637
CAS Number: 92952-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR